Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UTP14A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | UTP14A |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
UTP14A Polyclonal specifically detects UTP14A in Human samples. It is validated for Western Blot.Specifications
UTP14A | |
Polyclonal | |
Rabbit | |
Q9BVJ6 | |
10813 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
Antigen NY-CO-16, dJ537K23.3, FLJ37089, NYCO16, NY-CO-16, SDCCAG16, Serologically defined colon cancer antigen 16KIAA0266, U3 small nucleolar RNA-associated protein 14 homolog A, UTP14, U3 small nucleolar ribonucleoprotein, homolog A (yeast) | |
UTP14A | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title