Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UTP18 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB044207 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB044207 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB044207 Supplier Novus Biologicals Supplier No. NBP297492H

Rabbit Polyclonal Antibody

UTP18 Polyclonal specifically detects UTP18 in Human samples. It is validated for Western Blot.

Specifications

Antigen UTP18
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot
Formulation PBS
Gene Alias CGI-48, U3 small nucleolar RNA-associated protein 18 homolog, UTP18, small subunit (SSU) processome component, homolog (yeast), WD repeat domain 50, WD repeat-containing protein 50, WDR50
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human UTP18 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 51096
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.