Learn More
Honeywell™ Uvex™ Protege™ Safety Eyewear with Hydroshield™ Anti-Fog Lens Coating
Delivers anti-fog protection that lasts up to 60X longer
Specifications
Lens Type HydroShield AF
Frame Color Black
View More Specs
Products 4
Catalog Number Mfr. No. Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. Lens Tint Price Quantity & Availability  
19-018-529
View Documents
Honeywell™
S4201HS
Gray N/A N/A N/A
19-018-528
View Documents
Honeywell™
S4200HS
Clear N/A N/A N/A
19-185-713
View Documents
Honeywell™
S4222HS
Amber N/A N/A N/A
19-185-759
View Documents
Honeywell™
S4206HS
SCT-Low IR N/A N/A N/A
Description
Description
  • Provides 99.9% ultra-violet protection
  • Provides 2X greater scratch-resistance
Compliance
Meet ANSI™ Z87.1-2010 standards. Meet military V0 standard for impact protection
Specifications
Specifications
HydroShield AF
Black
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.