Learn More
Honeywell™ Uvex™ Livewire™ Sealed Eyewear
For protecting workers against intense wind, sun, dirt, dust, debris, as well as high heat and humidity
Specifications
For Use With (Application) | Agriculture, Construction, Forestry, Manufacturing, Mining, Municipal Services, Oil and Gas, Steel and Metals, Utilities, Welding, Wind Energy |
---|
Alert:
Not for chemical splash applications
Catalog Number | Mfr. No. | Lens Tint | Frame Color | Lens Coating | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Lens Tint | Frame Color | Lens Coating | Price | Quantity & Availability | |||
19-039-920
|
Honeywell™
S2600XP |
Clear | Matte Black | Anti-static, Scratch-resistant, UV-treated | N/A | N/A | N/A | ||
19-039-921
|
Honeywell™
S2600D |
Clear | Matte Black | Anti-fog, Scratch-resistant | N/A | N/A | N/A | ||
19-039-922
|
Honeywell™
S2601XP |
Espresso | Matte Black | Anti-static, Scratch-resistant, UV-treated | N/A | N/A | N/A | ||
19-039-923
|
Honeywell™
S2602XP |
Amber | Matte Black | Anti-static, Scratch-resistant, UV-treated | N/A | N/A | N/A | ||
19-039-924
|
Honeywell™
S2604XP |
SCT-Reflect 50 | Matte Black | Anti-static, Scratch-resistant, UV-treated | N/A | N/A | N/A | ||
19-039-925
|
Honeywell™
S2605XP |
SCT-Gray Lens | Matte Black | Anti-static, Scratch-resistant, UV-treated | N/A | N/A | N/A | ||
19-039-928
|
Honeywell™
S2609XP |
SCT-Low IR | Matte Black | Anti-static, Scratch-resistant, UV-treated | N/A | N/A | N/A | ||
19-039-929
|
Honeywell™
S2620XP |
Clear | Silver | Anti-static, Scratch-resistant, UV-treated | N/A | N/A | N/A | ||
19-039-930
|
Honeywell™
S2621XP |
Gray | Silver | Anti-static, Scratch-resistant, UV-treated | N/A | N/A | N/A | ||
19-039-931
|
Honeywell™
S2624XP |
SCT-Reflect 50 | Silver | Anti-static, Scratch-resistant, UV-treated | N/A | N/A | N/A | ||
19-185-763
|
Honeywell™
S2600HS |
Clear | Black | Anti-fog, Anti-static, Scratch-resistant, UV-treated | N/A | N/A | N/A | ||
Description
- Uvextreme Plus™ anti-fog lens coating with 10X longer-lasting anti-fog performance
- Also available with Dura-streme dual anti-fog/anti-scratch coating
- Foam-lined subframe
- Gap-free seal around eyes to protect from debris
- Flexible design can be worn with headband like goggle or with temples like spectacle
- Padded temples provide ideal tension and flex for comfort fit
- Ratcheting temples adjust lens angle by up to 15° for optimal coverage and air circulation
- Adjustable, flame-resistant latex-free cloth headband
- Wraparound frame design accommodates respirators, low-profile temples fits with earmuffs
- Wide lens tint assortment available in sealed eyewear: Clear, Amber, Espresso, Gray, SCT-Reflect 50 (indoor/outdoor mirror), SCT-Gray, SCT-Low IR, Shade 3.0 and Shade 5.0
- Available in Matte Black and Silver frame options
Certifications
CSA
Compliance
ANSI Z87.1-2010
Specifications
Agriculture, Construction, Forestry, Manufacturing, Mining, Municipal Services, Oil and Gas, Steel and Metals, Utilities, Welding, Wind Energy |