Learn More
Honeywell Safety Products™ Uvex™ Skyper™ Safety Glasses
Custom fit with superior peripheral protection
Specifications
Certifications/Compliance Meet ANSI Z87.1-2003 and CSA Z94.3-1999 standards.
Frame Color Black
Lens Material Polycarbonate
View More Specs
Products 4
Catalog Number Mfr. No. LensType Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. LensType Lens Tint Price Quantity & Availability  
19-038-311
View Documents
Honeywell Safety Products™
S1908
IR Filter Shade 5.0 N/A N/A N/A
19-038-310
View Documents
Honeywell Safety Products™
S1907
IR Filter Shade 3.0 N/A N/A N/A
19-038-309
View Documents
Honeywell Safety Products™
S1906
IR Filter Shade 2.0 N/A N/A N/A
19-037-656
Accessories View Documents
Honeywell Safety Products™
S1900
Clear N/A N/A N/A
Description
Description
Custom fit with superior peripheral protection
  • Polycarbonate lenses with single-lens design feature Ultradura™ hardcoat (UD) or Uvextreme™ AF (XTR) antifog, antiscratch, and antistatic coating
  • Wraparound, impact resistant lenses block 99.9% of harmful UVA and UVB radiation
  • Black frames with molded-in browguard reduce the danger of flying particulates
  • Molded-in nosebridge offers comfort and safety in a variety of work applications
  • Patented, adjustable, comfort-cushioned Duoflex™ temples
  • Simple lens replacement system, ratcheting lens inclination
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3-1999 standards.
Specifications
Specifications
Meet ANSI Z87.1-2003 and CSA Z94.3-1999 standards.
Black
Polycarbonate
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.