Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UVP Clear UV Blocking Covers

Catalog No. p-2621985 Shop All UVP Inc. Products
Click to view available options
For Use With (Equipment):
3uv™ transilluminators
Benchtop transilluminators
Standard Format UV Transilluminators

Specifications

For Use With (Equipment) Standard Format UV Transilluminators
For Use With (Application) Gel electrophoresis
Size 18 x 13 in.
Includes Uv Blocking Cover
Quantity each
Color Clear
Product Type UV Blocking Cover
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.