Learn More
Analytik Jena UVP eLITE
Catalog No. UVP98010501 Shop All UVP Inc. Products
Change view
Click to view available options
For Use With (Equipment):
UVP ChemStudio Systems
Catalog No. For Use With (Equipment)
UVP98010501 UVP ChemStudio Systems
1 options
Catalog No. UVP98010501
Supplier: Analytik Jena 98010501
Only null left
Add to Cart
Add to Cart
Request a Quote
For use with Analytik Jena US Advanced Imaging Systems.
The UVP eLITE Light Source uses Xenon light engine technology to provide uniform and high-intensity light across the full range of brilliant mulispectral excitation which can be produced for a variety of fluorescent dyes.
Specifications
For Use With (Equipment) UVP ChemStudio Systems
Voltage 115V