Learn More
UVP Firstlight UV Illuminator FI-26
Shop All UVP Inc. Products
Click to view available options
For Use With (Equipment):
Uvp Bioimaging System and CCD Camera
Description
Ideal source for quantitative fluorescent imaging in a wide range of genomic and proteomic applications
- Electrophoretic separation and quantitation of 1D and 2D protein separations
- Solid phase immunoassay
- DNA quantitation
- RNA quantitation
Specifications
Specifications
| For Use With (Equipment) | Uvp Bioimaging System and CCD Camera |
| For Use With (Application) | Used for low cost configurations with accurate gel to gel comparison |
| Includes | Uv blocking cover, uv transmitting gel-tray with cm markings, gel-cutter tool, fluorescent target |
| Wavelength | 302nm |
| Product Type | UV Illuminator |