Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Analytik Jena UVP Solo Elite Imaging System, 6.3 MPx

Catalog No. 84997094403
Click to view available options
Camera Type:
6.3 MP Color
6.3 MP Monochrome
Voltage:
115 V
230 V

The UVP Solo Elite is the perfect addition to any lab running routine gel analysis, imaging and documentation. This compact imager provides researchers rapid processing in a bench space saving format. With the most competitive specifications on the market and a budget friendly price, the UVP Solo Elite is the right fit for your lab.

  • This all-in-one system for gel documentation features an integrated 11.6 in. touch screen computer, a high resolution 6.3 MP camera - available as color or monochrome with an automated lens, a three position filter tray with pre-installed filters, overhead blue and white LED lighting and the UVP Elite UV Transilluminator.
  • Easily switch between emission filters
    • The three-position emission filter tray can be used to place three different filters and switch between them for different applications.
    • A broad band filter (535 nm to 660 nm) is included with the system and a variety of other emission filters are available for additional purchase.
  • Camera
    • Choose from a 6.3MP color or monochrome camera with an automated lens.
    • The Elite color camera can capture the natural color of the samples, eliminating the need for multiplexing and reducing post image processing for high quality color images.
  • Intuitive Software with easy access to capture controls
    • VisionWorks software on the UVP Solo Elite is a powerful yet intuitive software for capturing gel images and analyzing them.
    • The simple user interface enables easy access to auto and manual image capture modes under the Device tab. Exposure time can be adjusted with customizable exposure settings.
    • Captured images are saved in TIFF or JPEG formats.
  • Extensive image enhancement and analysis tools
    • Image enhancement and analysis features are included with all systems.
    • Researchers can personalize their experiments and make use of enhancement features and annotation tools for publication purposes.
    • The software offers many powerful tools such as noise reduction, histogram adjustment, inversion, pseudocolor, and more.
    • The extensive analysis features are optimized for accurate and reproducible results.
    • Several custom tools are available, such as automatic lane and band detection, 1D analysis, area density, and colony counting.
    • Once the quantitation results have been generated, reports are created to show thorough analysis, such as Molecular Weight (MW), Rf, band intensities, and area density calculations.
    • All the data can be conveniently exported to Excel.

Specifications

Voltage 115 V
Filter Type Ethidium Bromide
Resolution 3072 x 2048 pixel
Software VisionWorks
Transilluminator UVP Elite UV Transilluminator
Lens Type Automated
Camera Type 6.3 MP Color
Content And Storage 128 GB
Dimensions 61 x 46 x 38 cm
For Use With (Application) Fluorescent DNA Gels, Fluorescent Protein Gels, Blue Light Gels, Colorimetry and More
Color Black
Product Type Bioimaging
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.