Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UVRAG Antibody (953035), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB345836 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345836 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345836 Supplier Novus Biologicals Supplier No. FAB9180MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UVRAG Monoclonal specifically detects UVRAG in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen UVRAG
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 953035
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias beclin 1 binding protein, DHTX, disrupted in heterotaxy, p63, UV radiation resistance associated gene, UV radiation resistance-associated gene protein, VPS38
Host Species Mouse
Immunogen Human UVRAG synthetic peptide corresponding to sequences between amino acids 260-360, Accession # Q9P2Y5
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, Cellular Signaling, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 7405
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.