Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UXS1 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB181220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB181220 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB181220 Supplier Novus Biologicals Supplier No. NBP306482B
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UXS1 Polyclonal antibody specifically detects UXS1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen UXS1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias FLJ23591, SDR6E1, short chain dehydrogenase/reductase family 6E, member 12, UDP-glucuronate decarboxylase 1UXS-1, UDP-glucuronic acid decarboxylase 1, UGDEC 4.1.1.35
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human UXS1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 80146
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.