Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UXT Antibody, Novus Biologicals™
SDP

Catalog No. NBP100129 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP100129 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP100129 Supplier Novus Biologicals Supplier No. NBP100129
Add to Cart
Add to Cart

Goat Polyclonal Antibody

UXT Polyclonal specifically detects UXT in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen UXT
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.01 - 0.03 ug/ml, Immunohistochemistry 5 - 10 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml, Peptide ELISA Detection limit 1:64000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_004173.1
Gene Alias Androgen receptor trapped clone 27 protein, ART-27protein UXT, SKP2-associated alpha PFD 1, STAP1, Ubiquitously expressed transcript protein, ubiquitously-expressed transcript
Gene Symbols UXT
Host Species Goat
Immunogen Peptide with sequence C-DHRDKVYEQLAKY corresponding to C-Terminus according to NP_004173.1, NP_705582.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 8409
Test Specificity This antibody is expected to recognize both reported isoforms (NP_004173.1; NP_705582.1).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.