Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OTOS Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | OTOS |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19841720
![]() |
Novus Biologicals
NBP19841720UL |
20 μL |
Each for $206.00
|
|
|||||
NBP198417
![]() |
Novus Biologicals
NBP198417 |
100 μL |
Each for $487.50
|
|
|||||
Description
OTOS Polyclonal specifically detects OTOS in Mouse samples. It is validated for Western Blot.Specifications
OTOS | |
Polyclonal | |
Rabbit | |
NP_694754 | |
150677 | |
The immunogen for this antibody is Otos - middle region. Peptide sequence YWPFSTSDFWNYVQYFQTQGAYPQIEDMARTFFAHFPLGSTLGFHVPYQE. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
OTOSP, otospiralin | |
OTOS | |
IgG | |
10 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title