Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNCK Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP19842620UL
Description
PNCK Polyclonal specifically detects PNCK in Rat samples. It is validated for Western Blot.Specifications
PNCK | |
Polyclonal | |
Western Blot 1:1000 | |
NP_058971 | |
PNCK | |
The immunogen for this antibody is Pnck - C-terminal region. Peptide sequence QKNFARTHWKRAFNATSFLRHIRKLGQSPEGEEASRQGMTRHSHPGLGTS. | |
Affinity Purified | |
RUO | |
139728 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
BSTK3, calcium/calmodulin-dependent protein kinase type 1B, CaM kinase I beta, CaM kinase IB, CaMK1b, CaM-KI beta, CaMKI-beta, EC 2.7.11, EC 2.7.11.17, FLJ50403, FLJ50549, FLJ56451, FLJ59811, MGC45419, pregnancy up-regulated non-ubiquitously expressed CaM kinase, pregnancy upregulated non-ubiquitously expressed CaM kinase, Pregnancy up-regulated non-ubiquitously-expressed CaM kinase | |
Rabbit | |
37 kDa | |
20 μL | |
Primary | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction