Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNCK Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $499.50
Specifications
Antigen | PNCK |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19842620
![]() |
Novus Biologicals
NBP19842620UL |
20 μL |
Each for $158.00
|
|
|||||
NBP198426
![]() |
Novus Biologicals
NBP198426 |
100 μL |
Each for $499.50
|
|
|||||
Description
PNCK Polyclonal specifically detects PNCK in Rat samples. It is validated for Western Blot.Specifications
PNCK | |
Polyclonal | |
Rabbit | |
NP_058971 | |
139728 | |
The immunogen for this antibody is Pnck - C-terminal region. Peptide sequence QKNFARTHWKRAFNATSFLRHIRKLGQSPEGEEASRQGMTRHSHPGLGTS. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
BSTK3, calcium/calmodulin-dependent protein kinase type 1B, CaM kinase I beta, CaM kinase IB, CaMK1b, CaM-KI beta, CaMKI-beta, EC 2.7.11, EC 2.7.11.17, FLJ50403, FLJ50549, FLJ56451, FLJ59811, MGC45419, pregnancy up-regulated non-ubiquitously expressed CaM kinase, pregnancy upregulated non-ubiquitously expressed CaM kinase, Pregnancy up-regulated non-ubiquitously-expressed CaM kinase | |
PNCK | |
IgG | |
37 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title