Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SQRDL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP19843120UL
Description
SQRDL Polyclonal specifically detects SQRDL in Mouse samples. It is validated for Western Blot.Specifications
SQRDL | |
Polyclonal | |
Western Blot 1:1000 | |
NP_067482 | |
SQRDL | |
The immunogen for this antibody is Sqrdl - C-terminal region. Peptide sequence AQSGILDRTMCLIMKNQRPIKKYDGYTSCPLVTGYNRVILAEFDYTAQPL. | |
Affinity Purified | |
RUO | |
58472 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
CGI-44, sulfide dehydrogenase like, sulfide quinone reductase-like (yeast), sulfide:quinone oxidoreductase, mitochondrial | |
Rabbit | |
50 kDa | |
20 μL | |
Primary | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction