Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SQRDL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00
Specifications
Antigen | SQRDL |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP1984320
![]() |
Novus Biologicals
NBP19843120UL |
20 μL |
Each for $158.00
|
|
|||||
NBP198431
![]() |
Novus Biologicals
NBP198431 |
100 μL | N/A | N/A | N/A | ||||
Description
SQRDL Polyclonal specifically detects SQRDL in Mouse samples. It is validated for Western Blot.Specifications
SQRDL | |
Polyclonal | |
Rabbit | |
NP_067482 | |
58472 | |
The immunogen for this antibody is Sqrdl - C-terminal region. Peptide sequence AQSGILDRTMCLIMKNQRPIKKYDGYTSCPLVTGYNRVILAEFDYTAQPL. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
CGI-44, sulfide dehydrogenase like, sulfide quinone reductase-like (yeast), sulfide:quinone oxidoreductase, mitochondrial | |
SQRDL | |
IgG | |
50 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title