Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABLIM1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | ABLIM1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19847220
![]() |
Novus Biologicals
NBP19847220UL |
20 μL |
Each for $206.00
|
|
|||||
NBP198472
![]() |
Novus Biologicals
NBP198472 |
100 μL |
Each for $487.50
|
|
|||||
Description
ABLIM1 Polyclonal specifically detects ABLIM1 in Mouse samples. It is validated for Western Blot.Specifications
ABLIM1 | |
Polyclonal | |
Rabbit | |
NP_001096648 | |
3983 | |
The immunogen for this antibody is Ablim1 - middle region. Peptide sequence KAIYDIERPDLITYEPFYTSGYEDKQERQSLGESPRTLSPTPSAEGYQDV. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
ABLIM, abLIM-1, actin binding LIM protein 1, Actin-binding double zinc finger protein, actin-binding double-zinc-finger protein, actin-binding LIM protein 1, Actin-binding LIM protein family member 1, DKFZp781D0148, FLJ14564, KIAA0059, LIM actin-binding protein 1, LIMAB1MGC1224, LIMATIN | |
ABLIM1 | |
IgG | |
62 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title