Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ETEA Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP198272
Description
ETEA Polyclonal specifically detects ETEA in Mouse samples. It is validated for Western Blot.Specifications
ETEA | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
B17.2L, ETEAFAS-associated factor 2, expressed in T-cells and eosinophils in atopic dermatitis, Fas associated factor family member 2, KIAA0887UBX domain protein 3B, MMTN, NDUNDUFA12L, UBX domain containing 8, UBX domain-containing protein 3B, UBX domain-containing protein 8, UBXD8, UBXN3BProtein ETEA | |
Rabbit | |
Affinity purified | |
RUO | |
23197 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_848484 | |
FAF2 | |
The immunogen for this antibody is Faf2 antibody - N-terminal region. Peptide sequence LLQFQDLTGIESMEQCRLALEQHNWNMEAAVQDRLNEQEGVPSVFNPPPA. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 100%; Chicken: 100%; Canine: 100%; Guinea pig: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%; Xenopus: 92%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction