Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ETEA Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | ETEA |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ETEA Polyclonal specifically detects ETEA in Mouse samples. It is validated for Western Blot.Specifications
ETEA | |
Polyclonal | |
Rabbit | |
NP_848484 | |
23197 | |
The immunogen for this antibody is Faf2 antibody - N-terminal region. Peptide sequence LLQFQDLTGIESMEQCRLALEQHNWNMEAAVQDRLNEQEGVPSVFNPPPA. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
B17.2L, ETEAFAS-associated factor 2, expressed in T-cells and eosinophils in atopic dermatitis, Fas associated factor family member 2, KIAA0887UBX domain protein 3B, MMTN, NDUNDUFA12L, UBX domain containing 8, UBX domain-containing protein 3B, UBX domain-containing protein 8, UBXD8, UBXN3BProtein ETEA | |
FAF2 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title