Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RILP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$206.00 - $499.50
Specifications
Antigen | RILP |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19827820
![]() |
Novus Biologicals
NBP19827820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP198278
![]() |
Novus Biologicals
NBP198278 |
100 μL |
Each for $499.50
|
|
|||||
Description
RILP Polyclonal specifically detects RILP in Human samples. It is validated for Western Blot.Specifications
RILP | |
Polyclonal | |
Rabbit | |
Human | |
rab-interacting lysosomal protein, PP10141, Rab interacting lysosomal protein | |
RILP | |
IgG | |
44 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_113618 | |
83547 | |
The immunogen for this antibody is RILP antibody - C-terminal region. Peptide sequence PPPPESKIQSFFGLWYRGKAESSEDETSSPAPSKLGGEEEAQPQSPAPDP. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title