Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOS1AP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP19848720UL
Description
NOS1AP Polyclonal specifically detects NOS1AP in Human samples. It is validated for Western Blot.Specifications
| NOS1AP | |
| Polyclonal | |
| Western Blot 1:1000 | |
| BAA32309 | |
| NOS1AP | |
| The immunogen for this antibody is NOS1AP - C-terminal region. Peptide sequence PEDTPPPAQGEALLGGLELIKFRESGIASEYESNTDESEERDSWSQEELP. | |
| Affinity Purified | |
| RUO | |
| 9722 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| PBS and 2% Sucrose with 0.09% Sodium Azide | |
| 6330408P19Rik, CAPONMGC138500, carboxyl-terminal PDZ ligand of neuronal nitric oxide synthase protein, C-terminal PDZ ligand of neuronal nitric oxide synthase protein, KIAA0464C-terminal PDZ domain ligand of neuronal nitric oxide synthase (CAPON), ligand of neuronal nitric oxide synthase with carboxyl-terminal PDZ domain, nitric oxide synthase 1 (neuronal) adaptor protein, Nitric oxide synthase 1 adaptor protein | |
| Rabbit | |
| 63 kDa | |
| 20 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction