Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Snx6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP198504
Description
Snx6 Polyclonal specifically detects Snx6 in Human samples. It is validated for Western Blot.Specifications
| Snx6 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| MGC3157, MSTP010, sorting nexin 6, sorting nexin-6, TFAF2, TRAF4-associated factor 2, tumor necrosis factor receptor-associated factor 4(TRAF4)-associated factor 2 | |
| Rabbit | |
| 48 kDa | |
| 100 μL | |
| Signal Transduction | |
| 58533 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_689419 | |
| SNX6 | |
| The immunogen for this antibody is SNX6 - C-terminal region. Peptide sequence LRESQAAKDLLYRRSRSLVDYENANKALDKARAKNKDVLQAETSQQLCCQ. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction