Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Snx6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
| Antigen | Snx6 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19850420
![]() |
Novus Biologicals
NBP19850420UL |
20 μL |
Each for $206.00
|
|
|||||
NBP198504
![]() |
Novus Biologicals
NBP198504 |
100 μL |
Each for $487.50
|
|
|||||
Description
Snx6 Polyclonal specifically detects Snx6 in Human samples. It is validated for Western Blot.Specifications
| Snx6 | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| MGC3157, MSTP010, sorting nexin 6, sorting nexin-6, TFAF2, TRAF4-associated factor 2, tumor necrosis factor receptor-associated factor 4(TRAF4)-associated factor 2 | |
| SNX6 | |
| IgG | |
| 48 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| NP_689419 | |
| 58533 | |
| The immunogen for this antibody is SNX6 - C-terminal region. Peptide sequence LRESQAAKDLLYRRSRSLVDYENANKALDKARAKNKDVLQAETSQQLCCQ. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title