Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INO80B Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP19850620UL

 View more versions of this product

Catalog No. NBP19850620


Description

Description

INO80B Polyclonal specifically detects INO80B in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

INO80B
Polyclonal
Western Blot 1:1000
NP_076036
INO80B
The immunogen for this antibody is Ino80b - C-terminal region. Peptide sequence SPTLPLPVGGGCPAPALTEEMLLKREERARKRRLQAARRAEEHKNQTIER.
Affinity Purified
RUO
83444
Store at -20C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
HMGIYL4, IES2, IES2 homolog, INO80 complex subunit B, PAP-1 binding protein, PAP-1BP, PAPA1, zinc finger, HIT type 4
Rabbit
37 kDa
20 μL
Primary
Mouse
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.