Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cdc14A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | Cdc14A |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Cdc14A Polyclonal specifically detects Cdc14A in Human samples. It is validated for Western Blot.Specifications
| Cdc14A | |
| Polyclonal | |
| Rabbit | |
| Cell Cycle and Replication | |
| CDC10 (cell division cycle 10, S. cerevisiae, homolog), cdc14, CDC14 cell division cycle 14 homolog A, CDC14 cell division cycle 14 homolog A (S. cerevisiae), Cdc14A1, Cdc14A2, dual specificity protein phosphatase CDC14A, EC 3.1.3.16, EC 3.1.3.48, hCDC14 | |
| CDC14A | |
| IgG | |
| 66 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| NP_003663 | |
| 8556 | |
| The immunogen for this antibody is CDC14A - C-terminal region. Peptide sequence DDDVEMKNGITQGDKLRALKSQRQPRTSPSCAFRSDDTKGHPRAVSQPFR. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title