Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GNA14 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP198546
Description
GNA14 Polyclonal specifically detects GNA14 in Human samples. It is validated for Western Blot.Specifications
GNA14 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
G alpha-14, G-protein subunit alpha-14, guanine nucleotide binding protein (G protein), alpha 14, guanine nucleotide-binding protein 14, guanine nucleotide-binding protein subunit alpha-14 | |
Rabbit | |
41 kDa | |
100 μL | |
Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
9630 | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_004288 | |
GNA14 | |
The immunogen for this antibody is GNA14 - N-terminal region. Peptide sequence QLRRDKKDARRELKLLLLGTGESGKSTFIKQMRIIHGSGYSDEDRKGFTK. | |
Affinity purified | |
RUO | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction