Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRFAP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | MRFAP1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19855220
![]() |
Novus Biologicals
NBP19855220UL |
20 μL |
Each for $206.00
|
|
|||||
NBP198552
![]() |
Novus Biologicals
NBP198552 |
100 μL |
Each for $487.50
|
|
|||||
Description
MRFAP1 Polyclonal specifically detects MRFAP1 in Human samples. It is validated for Western Blot.Specifications
MRFAP1 | |
Polyclonal | |
Rabbit | |
NP_150638 | |
93621 | |
The immunogen for this antibody is MRFAP1 - N-terminal region. Peptide sequence PVINEMREDIASLTREHGRAYLRNRSKLWEMDNMLIQIKTQVEASEESAL. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Mof4 family associated protein 1, Morf4 family associated protein 1, PAM14MORF4 family-associated protein 1, PGR1protein associated with MRG, 14 kDa, Protein associated with MRG of 14 kDa, Protein PGR1, T-cell activation protein | |
MRFAP1 | |
IgG | |
15 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title