Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRFAP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP198552
Description
MRFAP1 Polyclonal specifically detects MRFAP1 in Human samples. It is validated for Western Blot.Specifications
MRFAP1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
Mof4 family associated protein 1, Morf4 family associated protein 1, PAM14MORF4 family-associated protein 1, PGR1protein associated with MRG, 14 kDa, Protein associated with MRG of 14 kDa, Protein PGR1, T-cell activation protein | |
Rabbit | |
15 kDa | |
100 μL | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_150638 | |
MRFAP1 | |
The immunogen for this antibody is MRFAP1 - N-terminal region. Peptide sequence PVINEMREDIASLTREHGRAYLRNRSKLWEMDNMLIQIKTQVEASEESAL. | |
Affinity purified | |
RUO | |
93621 | |
Human, Pig, Bovine, Canine, Rabbit | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MRFAP1 Antibody, Novus Biologicals™ > 100μL; Unlabeled
Spot an opportunity for improvement?Share a Content Correction