Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP4R4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP19856720UL
Description
PPP4R4 Polyclonal specifically detects PPP4R4 in Mouse samples. It is validated for Western Blot.Specifications
PPP4R4 | |
Polyclonal | |
Western Blot 1:1000 | |
NP_083256 | |
PPP4R4 | |
The immunogen for this antibody is Ppp4r4 - C-terminal region. Peptide sequence VKKTVLELDRMEMSMDMFQKKNYEKDLLDQEKEREELLFLEMEQLEKEKH. | |
Affinity Purified | |
RUO | |
57718 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
KIAA1622PP4R4HEAT-like repeat-containing protein, protein phosphatase 4, regulatory subunit 4, serine/threonine-protein phosphatase 4 regulatory subunit 4 | |
Rabbit | |
96 kDa | |
20 μL | |
Primary | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction