Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP4R4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | PPP4R4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP198567
![]() |
Novus Biologicals
NBP198567 |
100 μL |
Each for $487.50
|
|
|||||
NBP19856720
![]() |
Novus Biologicals
NBP19856720UL |
20 μL | N/A | N/A | N/A | ||||
Description
PPP4R4 Polyclonal specifically detects PPP4R4 in Mouse samples. It is validated for Western Blot.Specifications
PPP4R4 | |
Polyclonal | |
Rabbit | |
NP_083256 | |
57718 | |
The immunogen for this antibody is Ppp4r4 - C-terminal region. Peptide sequence VKKTVLELDRMEMSMDMFQKKNYEKDLLDQEKEREELLFLEMEQLEKEKH. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
KIAA1622PP4R4HEAT-like repeat-containing protein, protein phosphatase 4, regulatory subunit 4, serine/threonine-protein phosphatase 4 regulatory subunit 4 | |
PPP4R4 | |
IgG | |
96 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title