Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIP1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP198293 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP198293 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP198293 Supplier Novus Biologicals Supplier No. NBP198293
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRIP1 Polyclonal specifically detects TRIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen TRIP1
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_003748
Gene Alias eIF-3-beta, eIF3-beta, eIF3i, eIF3-p36, EIF3S2TGF-beta receptor-interacting protein 1, Eukaryotic translation initiation factor 3 subunit 2, eukaryotic translation initiation factor 3 subunit I, eukaryotic translation initiation factor 3, subunit 2 (beta, 36kD), eukaryotic translation initiation factor 3, subunit 2 beta, 36kDa, eukaryotic translation initiation factor 3, subunit I, predicted protein of HQ2242, TGFbeta receptor-interacting protein 1, TRIP-1, TRIP-1eIF3 p36, TRIP1PRO2242
Gene Symbols EIF3I
Host Species Rabbit
Immunogen The immunogen for this antibody is EIF3I - C-terminal region. Peptide sequence NVKEHSRQINDIQLSRDMTMFVTASKDNTAKLFDSTTLEHQKTFRTERPV.
Molecular Weight of Antigen 36 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8668
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.