Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TDRD7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP19858520UL
Description
TDRD7 Polyclonal specifically detects TDRD7 in Rat samples. It is validated for Western Blot.Specifications
TDRD7 | |
Polyclonal | |
Western Blot 1:1000 | |
NP_620226 | |
TDRD7 | |
The immunogen for this antibody is Tdrd7 - C-terminal region. Peptide sequence CSDCSIKVTKVDEARGVAYVYLFTPKNFPDPHRSINRQITNADLWKHQKD. | |
Affinity Purified | |
RUO | |
23424 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
KIAA1529, PCTAIRE2-binding protein, PCTAIRE2BPTRAP, RP11-508D10.1, Trap, tudor domain containing 7, tudor domain-containing protein 7, Tudor repeat associator with PCTAIRE 2 | |
Rabbit | |
122 kDa | |
20 μL | |
Primary | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction