Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TDRD7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | TDRD7 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19858520
![]() |
Novus Biologicals
NBP19858520UL |
20 μL |
Each for $206.00
|
|
|||||
NBP198585
![]() |
Novus Biologicals
NBP198585 |
100 μL |
Each for $487.50
|
|
|||||
Description
TDRD7 Polyclonal specifically detects TDRD7 in Rat samples. It is validated for Western Blot.Specifications
TDRD7 | |
Polyclonal | |
Rabbit | |
NP_620226 | |
23424 | |
The immunogen for this antibody is Tdrd7 - C-terminal region. Peptide sequence CSDCSIKVTKVDEARGVAYVYLFTPKNFPDPHRSINRQITNADLWKHQKD. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
KIAA1529, PCTAIRE2-binding protein, PCTAIRE2BPTRAP, RP11-508D10.1, Trap, tudor domain containing 7, tudor domain-containing protein 7, Tudor repeat associator with PCTAIRE 2 | |
TDRD7 | |
IgG | |
122 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title