Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CKS1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | CKS1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP198307
![]() |
Novus Biologicals
NBP198307 |
100 μL |
Each for $487.50
|
|
|||||
NBP19830720
![]() |
Novus Biologicals
NBP19830720UL |
20 μL | N/A | N/A | N/A | ||||
Description
CKS1 Polyclonal specifically detects CKS1 in Mouse samples. It is validated for Western Blot.Specifications
CKS1 | |
Polyclonal | |
Rabbit | |
NP_058600 | |
1163 | |
The immunogen for this antibody is Cks1 - N-terminal region. Peptide sequence EFEYRHVMLPKDIAKLVPKTHLMSESEWRNLGVQQSQGWVHYMIHEPEPH. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
CDC28 protein kinase 1, CDC28 protein kinase 1B, CDC28 protein kinase regulatory subunit 1B, cell division control protein CKS1, CKS-1, CKS1CDC2-associated protein CKS1, ckshs1, cyclin-dependent kinases regulatory subunit 1, NB4 apoptosis/differentiation related protein, PNAS-143, PNAS-16, PNAS-18 | |
CKS1B | |
IgG | |
10 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title