Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF667 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP198333
Description
ZNF667 Polyclonal specifically detects ZNF667 in Rat samples. It is validated for Western Blot.Specifications
ZNF667 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
DKFZp686O111, FLJ14011, FLJ45518, MIPU1, myocardial ischemic preconditioning upregulated 1 ortholog, zinc finger protein 667 | |
Rabbit | |
66 kDa | |
100 μL | |
Primary | |
Porcine: 86%; Mouse: 86%; Equine: 85%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Rabbit | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_001008557 | |
ZNF667 | |
The immunogen for this antibody is Znf667 - N-terminal region. Peptide sequence MPAARGKSKSKAPVTFGDLAIYFSQEEWEWLSPNQKDLYEDVMLENYHNL. | |
Affinity purified | |
RUO | |
63934 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction