Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPRE1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP19836620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP19836620 20 μL
NBP198366 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP19836620 Supplier Novus Biologicals Supplier No. NBP19836620UL

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

MAPRE1 Polyclonal specifically detects MAPRE1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen MAPRE1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_031922
Gene Alias adenomatous polyposis coli-binding protein EB1, APC-binding protein EB1, EB1MGC129946, End-binding protein 1, MGC117374, microtubule-associated protein RP/EB family member 1, microtubule-associated protein, RP/EB family, member 1
Gene Symbols MAPRE1
Host Species Rabbit
Immunogen The immunogen for this antibody is Mapre1 - C-terminal region. Peptide sequence GKLRNIELICQENEGENDPVLQRIVDILYATDEGFVIPDEGGPQEEQEEY.
Molecular Weight of Antigen 30 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22919
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.