Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPRT Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | HPRT |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19835920
![]() |
Novus Biologicals
NBP19835920UL |
20 μL |
Each for $206.00
|
|
|||||
NBP198359
![]() |
Novus Biologicals
NBP198359 |
100 μL |
Each for $487.50
|
|
|||||
Description
HPRT Polyclonal specifically detects HPRT in Mouse samples. It is validated for Western Blot.Specifications
HPRT | |
Western Blot | |
Unconjugated | |
RUO | |
EC 2.4.2.8, HGPRTHPRTHGPRTase, hypoxanthine phosphoribosyltransferase 1, hypoxanthine-guanine phosphoribosyltransferase | |
HPRT1 | |
IgG | |
24 kDa |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Rabbit | |
P00493 | |
3251 | |
Peptide sequence (SRSVGYRPDFVGFEIPDKFVVGYALDYNEYFRDLNHVCVISETGKAKYKA) corresponding to aa 169-218 from C-terminal region of mouse HPRT (NP_038584). | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title