Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KERA Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP19838920UL
Description
KERA Polyclonal specifically detects KERA in Mouse samples. It is validated for Western Blot.Specifications
KERA | |
Polyclonal | |
Western Blot 1:1000 | |
NP_032464 | |
KERA | |
The immunogen for this antibody is Kera - C-terminal region. Peptide sequence MQLNMAKNALRNMPPRLPANTMQLFLDNNSIEGIPENYFNVIPKVAFLRL. | |
Affinity Purified | |
RUO | |
11081 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
CNA2, keratocan, KTN, SLRR2BKeratan sulfate proteoglycan keratocan | |
Rabbit | |
39 kDa | |
20 μL | |
Primary | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction