Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KERA Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $487.50
Specifications
| Antigen | KERA |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19838920
![]() |
Novus Biologicals
NBP19838920UL |
20 μL |
Each for $158.00
|
|
|||||
NBP198389
![]() |
Novus Biologicals
NBP198389 |
100 μL |
Each for $487.50
|
|
|||||
Description
KERA Polyclonal specifically detects KERA in Mouse samples. It is validated for Western Blot.Specifications
| KERA | |
| Polyclonal | |
| Rabbit | |
| NP_032464 | |
| 11081 | |
| The immunogen for this antibody is Kera - C-terminal region. Peptide sequence MQLNMAKNALRNMPPRLPANTMQLFLDNNSIEGIPENYFNVIPKVAFLRL. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| CNA2, keratocan, KTN, SLRR2BKeratan sulfate proteoglycan keratocan | |
| KERA | |
| IgG | |
| 39 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title