Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPS5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP19840320UL
Description
RPS5 Polyclonal specifically detects RPS5 in Human samples. It is validated for Western Blot.Specifications
| RPS5 | |
| Polyclonal | |
| Western Blot 1:1000 | |
| NP_001000 | |
| RPS5 | |
| The immunogen for this antibody is RPS5 - C-terminal region. Peptide sequence: LCTGAREAAFRNIKTIAECLADELINAAKGSSNSYAIKKKDELERVAKSN | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS and 2% Sucrose with 0.09% Sodium Azide | |
| ribosomal protein S5,40S ribosomal protein S5 | |
| Rabbit | |
| 23 kDa | |
| 20 μL | |
| Cancer, Core ESC Like Genes, Stem Cell Markers | |
| 6193 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction