Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPS5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | RPS5 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP198403
![]() |
Novus Biologicals
NBP198403 |
100 μL |
Each for $487.50
|
|
|||||
NBP19840320
![]() |
Novus Biologicals
NBP19840320UL |
20 μL | N/A | N/A | N/A | ||||
Description
RPS5 Polyclonal specifically detects RPS5 in Human samples. It is validated for Western Blot.Specifications
| RPS5 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Core ESC Like Genes, Stem Cell Markers | |
| ribosomal protein S5,40S ribosomal protein S5 | |
| RPS5 | |
| IgG | |
| 23 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| NP_001000 | |
| 6193 | |
| The immunogen for this antibody is RPS5 - C-terminal region. Peptide sequence: LCTGAREAAFRNIKTIAECLADELINAAKGSSNSYAIKKKDELERVAKSN | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title