Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eIF3K Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | eIF3K |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
eIF3K Polyclonal specifically detects eIF3K in Mouse samples. It is validated for Western Blot.Specifications
eIF3K | |
Polyclonal | |
Rabbit | |
NP_082935 | |
27335 | |
The immunogen for this antibody is Eif3k - N-terminal region. Peptide sequence QILLKALTNLPHTDFTLCKCMIDQAHQEERPIRQILYLGDLLETCHFQAF. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
ARG134, eIF3k, EIF3-p28, EIF3S12eIF-3 p28, Eukaryotic translation initiation factor 3 subunit 12, eukaryotic translation initiation factor 3 subunit K, eukaryotic translation initiation factor 3, subunit 12, eukaryotic translation initiation factor 3, subunit K, HSPC029, M9, MSTP001, muscle specific, Muscle-specific gene M9 protein, PLAC24, PLAC-24, PLAC-24eIF-3 p25, PRO1474, PTD001 | |
EIF3K | |
IgG | |
25 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title