Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V-type proton ATPase subunit F Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB080971 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB080971 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB080971 Supplier Novus Biologicals Supplier No. NBP297084AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

V-type proton ATPase subunit F Polyclonal specifically detects V-type proton ATPase subunit F in Human samples. It is validated for ELISA.

Specifications

Antigen V-type proton ATPase subunit F
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias ATP6S14adenosinetriphosphatase 14k chain, ATPase, H+ transporting, lysosomal 14kDa, V1 subunit F, H(+)-transporting two-sector ATPase, 14kD subunit, MGC117321, MGC126037, MGC126038, vacuolar ATP synthase subunit F, vacuolar proton pump F subunit, Vacuolar proton pump subunit F, VATFATPase, vacuolar, 14 kD, V-ATPase 14 kDa subunit, V-ATPase F subunit, V-ATPase subunit F, Vma7, V-type proton ATPase subunit F
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant V-type proton ATPase subunit F (NP_004222.2; Met1-Arg119).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 9296
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.