Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V-type proton ATPase subunit F Antibody, Novus Biologicals™
SDP

Catalog No. NB436166 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB436166 25 μL
NBP238587 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB436166 Supplier Novus Biologicals Supplier No. NBP23858725UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

V-type proton ATPase subunit F Polyclonal specifically detects V-type proton ATPase subunit F in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen V-type proton ATPase subunit F
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q16864
Gene Alias ATP6S14adenosinetriphosphatase 14k chain, ATPase, H+ transporting, lysosomal 14kDa, V1 subunit F, H(+)-transporting two-sector ATPase, 14kD subunit, MGC117321, MGC126037, MGC126038, vacuolar ATP synthase subunit F, vacuolar proton pump F subunit, Vacuolar proton pump subunit F, VATFATPase, vacuolar, 14 kD, V-ATPase 14 kDa subunit, V-ATPase F subunit, V-ATPase subunit F, Vma7, V-type proton ATPase subunit F
Gene Symbols ATP6V1F
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: DTFRSLGSLPGSVVEANPNQRDPPLWDEIDSRQFL
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 9296
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.