Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V1a Vasopressin R/AVPR1A Antibody (612130), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB305422 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305422 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB305422 Supplier Novus Biologicals Supplier No. FAB10932I
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

V1a Vasopressin R/AVPR1A Monoclonal specifically detects V1a Vasopressin R/AVPR1A in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen V1a Vasopressin R/AVPR1A
Applications Flow Cytometry
Classification Monoclonal
Clone 612130
Conjugate Janelia Fluor 549
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias Antidiuretic hormone receptor 1a, arginine vasopressin receptor 1A, AVPR V1a, AVPR1, SCCL vasopressin subtype 1a receptor, V1a vasopressin receptor, V1aR, V1-vascular vasopressin receptor AVPR1A, Vascular/hepatic-type arginine vasopressin receptor, vasopressin V1a receptor
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human V1a Vasopressin R/AVPR1A, Accession # P37288
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 552
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.