Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V2 Vasopressin R/AVPR2 Antibody, Novus Biologicals™
SDP

Catalog No. NB036657 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036657 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036657 Supplier Novus Biologicals Supplier No. NBP289072

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

V2 Vasopressin R/AVPR2 Polyclonal specifically detects V2 Vasopressin R/AVPR2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen V2 Vasopressin R/AVPR2
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS containing 50% glycerol
Gene Alias ADHRMGC138386, Antidiuretic hormone receptor, arginine vasopressin receptor 2, AVPR V2, DI1, DIR3, DIRNDI, Renal-type arginine vasopressin receptor, V2RMGC126533, vasopressin V2 receptor
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the sequence near the N-terminus of rat V2 Vasopressin R/AVPR2.
Purification Method Peptide affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cellular Markers, GPCR
Primary or Secondary Primary
Gene ID (Entrez) 554
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.