Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V5 Epitope Tag Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB60381A750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB60381A750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB60381A750 Supplier Novus Biologicals Supplier No. NB600381AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

V5 Epitope Tag Polyclonal antibody specifically detects V5 Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.

Specifications

Antigen V5 Epitope Tag
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias V5 epitope tag
Host Species Rabbit
Immunogen Rabbits were immunized with a synthetic peptide representing amino acid residues 95 to 108 of RNA polymerase alpha subunit of simion virus 5 conjugated to KLH.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Content And Storage Store at 4°C in the dark.
Isotype IgG
Target Molecule Epitope Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.