Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V5 Epitope Tag Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB600381B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600381B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600381B Supplier Novus Biologicals Supplier No. NB600381B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

V5 Epitope Tag Polyclonal antibody specifically detects V5 Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA.

Specifications

Antigen V5 Epitope Tag
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Biotin
Dilution Western Blot 1:10000-1:100000, ELISA 1:100-1:2000
Formulation PBS and 0.2% BSA with 0.05% Sodium Azide
Gene Alias V5 epitope tag
Host Species Rabbit
Immunogen Rabbits were immunized with a synthetic peptide representing amino acid residues 95 to 108 of RNA polymerase alpha subunit of simion virus 5 conjugated to KLH.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epitope Tags, V5 Epitope Tags
Primary or Secondary Primary
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Target Molecule Epitope Tag
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.