Learn More
V5 Epitope Tag Antibody (E10/V4RR), Alexa Fluor™ 532, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | V5 Epitope Tag |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | E10/V4RR |
| Conjugate | Alexa Fluor 532 |
| Dilution | Western Blot, ELISA, Immunofluorescence |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | V5 epitope tag |
| Host Species | Mouse |
| Immunogen | GKPIPNPLLGLDST (V5) synthetic peptide conjugated to KLH |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.